Assay Details
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106901.2 | NP_001100371.2 | 7 - 8 | 2109 | 72 | |
XM_006244756.2 | XP_006244818.1 | 7 - 8 | 2116 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
