Assay Details
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133392.1 | NP_596883.1 | 4 - 5 | 641 | 142 | |
XM_006244908.2 | XP_006244970.1 | 4 - 5 | 1410 | 142 | ||
GenBank mRNA | AB070349.1 | - | 4 - 5 | 641 | 142 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
