Assay Details
Gene Symbol: Nck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108216.1 | NP_001101686.1 | 4 - 5 | 1219 | 100 | |
XM_006244825.2 | XP_006244887.1 | 4 - 5 | 1184 | 100 | ||
XM_008767035.1 | XP_008765257.1 | 4 - 5 | 3628 | 100 | ||
GenBank mRNA | AY724492.1 | - | 4 - 5 | 1219 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
immunological synapse formation
actin filament organization
phosphorylation
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
negative regulation of peptidyl-serine phosphorylation
positive regulation of T cell proliferation
positive regulation of transcription by RNA polymerase II
ephrin receptor signaling pathway
regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
dendritic spine development
kinase activity
protein-containing complex binding
scaffold protein binding
immunological synapse formation
actin filament organization
phosphorylation
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
negative regulation of peptidyl-serine phosphorylation
positive regulation of T cell proliferation
positive regulation of transcription by RNA polymerase II
ephrin receptor signaling pathway
regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
dendritic spine development
kinase activity
protein-containing complex binding
scaffold protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P698520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27385 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P698520 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P698520 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P698520 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P207215 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78199 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59112 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75641 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68813 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10922961 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922958 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922960 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922957 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922959 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10922956 | - | - |
Rat Genome 230 2.0 Array | 1398441_at | - | - |
Rat Genome 230 2.0 Array | 1390056_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398441_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390056_at | - | - |
Affymetrix HT Rat Focus Array | 1398441_at | - | - |
Affymetrix HT Rat Focus Array | 1398441_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900004599.rn.1 | TC0900000574.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900004598.rn.1 | TC0900000574.rn.1 | RTA TC |
Rat Expression Set 230 | 1390056_at | - | - |
Rat Expression Set 230 | 1398441_PM_at | - | - |
Rat Expression Set 230 | 1390056_PM_at | - | - |
Rat Expression Set 230 | 1398441_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922956 | - | - |
Rat Genome U34 Set | rc_AA957410_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21078279 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359798 | - | - |
