Assay Details
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031510.1 | NP_113698.1 | 4 - 5 | 730 | 118 | |
XM_006245049.2 | XP_006245111.1 | 5 - 6 | 788 | 118 | ||
XM_008767076.1 | XP_008765298.1 | 5 - 6 | 820 | 118 | ||
GenBank mRNA | DQ403076.1 | - | 4 - 5 | 620 | 118 | |
FQ214242.1 | - | 5 - 6 | 811 | 118 | ||
L35317.1 | - | 4 - 5 | 730 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
