Assay Details
Gene Symbol: Aox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008522.1 | NP_001008522.1 | 13 - 14 | 1271 | 109 | |
XM_008767106.1 | XP_008765328.1 | 14 - 15 | 2900 | 109 | ||
GenBank mRNA | AY665588.1 | - | 13 - 14 | 1271 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
