Assay Details
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007742.1 | NP_001007743.1 | 1 - 2 | 307 | 106 | |
XM_006233689.1 | XP_006233751.1 | 1 - 2 | 334 | 106 | ||
GenBank mRNA | BC083603.1 | - | 1 - 2 | 307 | 7133164 | 106 |
FQ210767.1 | - | 1 - 2 | 193 | 106 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
