Assay Details
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007742.1 | NP_001007743.1 | 5 - 6 | 715 | 78 | |
XM_006233689.1 | XP_006233751.1 | 5 - 6 | 742 | 78 | ||
GenBank mRNA | BC083603.1 | - | 5 - 6 | 715 | 78 | |
FQ210767.1 | - | 5 - 6 | 601 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
