Assay Details
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007742.1 | NP_001007743.1 | 6 - 7 | 815 | 96 | |
XM_006233689.1 | XP_006233751.1 | 6 - 7 | 842 | 96 | ||
GenBank mRNA | BC083603.1 | - | 6 - 7 | 815 | 96 | |
FQ216687.1 | - | 1 - 2 | 50 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
