Assay Details
Gene Symbol: Med27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106565.1 | NP_001100035.1 | 3 - 4 | 517 | 122 | |
XM_008761638.1 | XP_008759860.1 | 3 - 4 | 523 | 122 | ||
XM_008761639.1 | XP_008759861.1 | 2 - 3 | 556 | 122 | ||
GenBank mRNA | BC167032.1 | - | 3 - 4 | 492 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
