Assay Details
Gene Symbol: Med27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106565.1 | NP_001100035.1 | 7 - 8 | 847 | 71 | |
XM_008761639.1 | XP_008759861.1 | 6 - 7 | 886 | 71 | ||
GenBank mRNA | BC167032.1 | - | 7 - 8 | 822 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2466 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14866 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14866 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14866 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14866 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47500 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97339 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7985 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25762 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10834944 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834939 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834943 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834938 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834936 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834937 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834941 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834942 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10834935 | - | - |
Rat Genome 230 2.0 Array | 1372014_at | - | - |
Rat Genome 230 2.0 Array | 1372014_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372014_at | - | - |
Affymetrix HT Rat Focus Array | 1372014_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300001821.rn.1 | TC0300000180.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300001822.rn.1 | TC0300000180.rn.1 | RTA TC |
Rat Expression Set 230 | 1372014_PM_at | - | - |
Rat Expression Set 230 | 1372014_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834935 | - | - |
Rat Genome U34 Set | rc_AA963072_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21600828 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359059 | - | - |
