Assay Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271240.1 | NP_001258169.1 | 1 - 2 | 197 | 74 | |
XM_006233999.2 | XP_006234061.1 | 4 - 5 | 554 | 74 | ||
XM_006234000.2 | XP_006234062.1 | 4 - 5 | 469 | 74 | ||
GenBank mRNA | BC158696.1 | - | 4 - 5 | 341 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
