Assay Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271240.1 | NP_001258169.1 | 2 - 3 | 285 | 90 | |
XM_006233999.2 | XP_006234061.1 | 5 - 6 | 642 | 90 | ||
XM_006234000.2 | XP_006234062.1 | 5 - 6 | 557 | 90 | ||
GenBank mRNA | BC158696.1 | - | 5 - 6 | 429 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
