Assay Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271240.1 | NP_001258169.1 | 6 - 7 | 1090 | 84 | |
XM_006233999.2 | XP_006234061.1 | 9 - 10 | 1447 | 84 | ||
XM_006234000.2 | XP_006234062.1 | 9 - 10 | 1362 | 84 | ||
GenBank mRNA | BC158696.1 | - | 9 - 10 | 1234 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
