Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013081.1 | NP_001013099.1 | 10 - 11 | 877 | 61 | |
XM_006233777.2 | XP_006233839.1 | 11 - 12 | 1261 | 61 | ||
XM_006233779.2 | XP_006233841.1 | 11 - 12 | 1954 | 61 | ||
XM_006233780.2 | XP_006233842.1 | 11 - 12 | 1017 | 61 | ||
GenBank mRNA | BC091389.1 | - | 10 - 11 | 877 | 7321523 | 61 |
FQ212963.1 | - | 10 - 11 | 893 | 61 | ||
FQ219402.1 | - | 10 - 11 | 898 | 61 | ||
FQ219572.1 | - | 7 - 8 | 880 | 61 | ||
FQ223356.1 | - | 10 - 11 | 884 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
