Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013081.1 | NP_001013099.1 | 7 - 8 | 496 | 101 | |
XM_006233777.2 | XP_006233839.1 | 8 - 9 | 880 | 101 | ||
XM_006233778.2 | XP_006233840.1 | 8 - 9 | 880 | 101 | ||
XM_006233779.2 | XP_006233841.1 | 8 - 9 | 1573 | 101 | ||
XM_006233780.2 | XP_006233842.1 | 8 - 9 | 636 | 101 | ||
GenBank mRNA | BC091389.1 | - | 7 - 8 | 496 | 101 | |
FQ212963.1 | - | 7 - 8 | 512 | 101 | ||
FQ219402.1 | - | 7 - 8 | 517 | 101 | ||
FQ219572.1 | - | 4 - 5 | 499 | 101 | ||
FQ223356.1 | - | 7 - 8 | 503 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
