Assay Details
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106567.1 | NP_001100037.1 | 3 - 4 | 462 | 71 | |
XM_006233783.2 | XP_006233845.1 | 3 - 4 | 1499 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
