Assay Details
Gene Symbol: Gpr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153295.1 | NP_695207.1 | 7 - 8 | 931 | 139 | |
GenBank mRNA | AF488741.1 | - | 7 - 8 | 931 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
