Assay Details
Gene Symbol: Gabrr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138897.1 | NP_620252.1 | 8 - 9 | 1289 | 75 | |
GenBank mRNA | D50671.1 | - | 8 - 9 | 1289 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136485 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164520 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164520 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164520 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13328 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22030 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19483 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19355 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10753544 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753548 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753549 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753550 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753547 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753542 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753543 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753545 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753546 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10753541 | - | - |
Rat Genome 230 2.0 Array | 1370702_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370702_at | - | - |
Affymetrix HT Rat Focus Array | 1370702_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370702_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100008708.rn.1 | TC1100001434.rn.1 | RTA TC |
Rat Expression Set 230 | 1370702_PM_at | - | - |
Rat Expression Set 230 | 1370702_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753541 | - | - |
Rat Genome U34 Set | D50671_at | - | - |
Rat Genome U34 Set | D50671_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22241617 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372968 | - | - |
RatRef-12 Expression BeadChip | ILMN_60642 | - | - |
