Assay Details
Gene Symbol: Cd86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020081.1 | NP_064466.1 | 4 - 5 | 705 | 154 | |
XM_006248399.2 | XP_006248461.1 | 4 - 5 | 1072 | 154 | ||
XM_006248400.1 | XP_006248462.1 | 3 - 4 | 681 | 154 | ||
GenBank mRNA | D50558.1 | - | 4 - 5 | 705 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
