Assay Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107923.2 | NP_001101393.1 | 14 - 15 | 1622 | 164 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
7-methylguanosine mRNA capping
RNA processing
biological_process
peptidyl-tyrosine dephosphorylation
polynucleotide 5' dephosphorylation
molecular_function
mRNA guanylyltransferase activity
polynucleotide 5'-phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
RNA guanylyltransferase activity
triphosphatase activity
7-methylguanosine mRNA capping
RNA processing
biological_process
peptidyl-tyrosine dephosphorylation
polynucleotide 5' dephosphorylation
molecular_function
mRNA guanylyltransferase activity
polynucleotide 5'-phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
RNA guanylyltransferase activity
triphosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P746068 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P319087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36287 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P319087 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P746068 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P746068 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P319087 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P319087 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P746068 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P319087 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76219 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69521 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81308 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32403 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98776 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10868180 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868184 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868177 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868181 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868173 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868178 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868174 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868183 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868176 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868185 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868172 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868182 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868175 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868179 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10868171 | - | - |
Rat Genome 230 2.0 Array | 1390475_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391921_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390475_at | - | - |
Rat Genome 230 2.0 Array | 1391921_at | - | - |
Affymetrix HT Rat Focus Array | 1391921_at | - | - |
Affymetrix HT Rat Focus Array | 1390475_at | - | - |
Affymetrix HT Rat Focus Array | 1391921_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390475_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500002564.rn.1 | TC0500000407.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002566.rn.1 | TC0500000407.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002565.rn.1 | TC0500000407.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002561.rn.1 | TC0500000407.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002567.rn.1 | TC0500000407.rn.1 | RTA TC |
Rat Expression Set 230 | 1390475_PM_at | - | - |
Rat Expression Set 230 | 1390475_at | - | - |
Rat Expression Set 230 | 1391921_PM_at | - | - |
Rat Expression Set 230 | 1391921_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868171 | - | - |
Rat Genome U34 Set | rc_AI227688_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20922953 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357672 | - | - |
