Assay Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107923.2 | NP_001101393.1 | 4 - 5 | 474 | 154 | |
XM_006237990.2 | XP_006238052.1 | 4 - 5 | 488 | 154 | ||
XM_006237991.2 | XP_006238053.1 | 4 - 5 | 460 | 154 | ||
GenBank mRNA | BC168990.1 | - | 4 - 5 | 470 | 8364610 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
