Assay Details
Gene Symbol: Aqp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031703.1 | NP_113891.1 | 2 - 3 | 306 | 90 | |
GenBank mRNA | BC127490.1 | - | 2 - 3 | 288 | 90 | |
D17695.1 | - | 2 - 3 | 221 | 90 | ||
L35108.1 | - | 2 - 3 | 306 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
