Assay Details
Gene Symbol: Slc35a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107924.1 | NP_001101394.1 | 4 - 5 | 562 | 92 | |
XM_006237996.2 | XP_006238058.1 | 5 - 6 | 745 | 92 | ||
GenBank mRNA | FQ213102.1 | - | 5 - 6 | 596 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
