Assay Details
Gene Symbol: Slc35a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107924.1 | NP_001101394.1 | 6 - 7 | 876 | 119 | |
XM_006237996.2 | XP_006238058.1 | 7 - 8 | 1059 | 119 | ||
GenBank mRNA | FQ213102.1 | - | 7 - 8 | 910 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P096472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130499 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096472 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096472 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P096472 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P544436 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10875908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875902 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875904 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875906 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875903 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875905 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875907 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10875901 | - | - |
Rat Genome 230 2.0 Array | 1386078_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374662_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375254_at | - | - |
Rat Genome 230 2.0 Array | 1386077_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386078_at | - | - |
Rat Genome 230 2.0 Array | 1386077_at | - | - |
Rat Genome 230 2.0 Array | 1375254_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374662_at | - | - |
Affymetrix HT Rat Focus Array | 1375254_at | - | - |
Affymetrix HT Rat Focus Array | 1386077_at | - | - |
Affymetrix HT Rat Focus Array | 1386078_at | - | - |
Affymetrix HT Rat Focus Array | 1374662_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386077_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374662_at | - | - |
Affymetrix HT Rat Focus Array | 1386078_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375254_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500018435.rn.1 | TC0500002495.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018436.rn.1 | TC0500002495.rn.1 | RTA TC |
Rat Expression Set 230 | 1374662_PM_at | - | - |
Rat Expression Set 230 | 1375254_PM_at | - | - |
Rat Expression Set 230 | 1386077_PM_at | - | - |
Rat Expression Set 230 | 1386077_at | - | - |
Rat Expression Set 230 | 1386078_PM_at | - | - |
Rat Expression Set 230 | 1375254_at | - | - |
Rat Expression Set 230 | 1374662_at | - | - |
Rat Expression Set 230 | 1386078_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875901 | - | - |
Rat Genome U34 Set | rc_AA891822_at | - | - |
Rat Genome U34 Set | rc_AA891848_at | - | - |
Rat Genome U34 Set | rc_AI072449_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22388639 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356217 | - | - |
