Assay Details
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301235.1 | NP_001288164.1 | 6 - 7 | 807 | 128 | |
XM_008763680.1 | XP_008761902.1 | 6 - 7 | 1147 | 128 | ||
XM_008763682.1 | XP_008761904.1 | 5 - 6 | 525 | 128 | ||
GenBank mRNA | FQ214609.1 | - | 6 - 7 | 742 | 128 | |
FQ214635.1 | - | 6 - 7 | 744 | 128 | ||
FQ214711.1 | - | 6 - 7 | 744 | 128 | ||
FQ214816.1 | - | 6 - 7 | 723 | 128 | ||
FQ214823.1 | - | 6 - 7 | 744 | 128 | ||
FQ214899.1 | - | 6 - 7 | 744 | 128 | ||
FQ215016.1 | - | 6 - 7 | 707 | 128 | ||
FQ215066.1 | - | 6 - 7 | 743 | 128 | ||
FQ215076.1 | - | 6 - 7 | 704 | 128 | ||
FQ215143.1 | - | 6 - 7 | 744 | 128 | ||
FQ215210.1 | - | 6 - 7 | 745 | 128 | ||
FQ215302.1 | - | 6 - 7 | 744 | 128 | ||
FQ215335.1 | - | 6 - 7 | 704 | 128 | ||
FQ215601.1 | - | 6 - 7 | 745 | 128 | ||
FQ215676.1 | - | 6 - 7 | 744 | 128 | ||
FQ215883.1 | - | 6 - 7 | 742 | 128 | ||
FQ215920.1 | - | 6 - 7 | 742 | 128 | ||
FQ216112.1 | - | 6 - 7 | 128 | |||
FQ216228.1 | - | 6 - 7 | 744 | 128 | ||
FQ216294.1 | - | 6 - 7 | 743 | 128 | ||
FQ216477.1 | - | 6 - 7 | 744 | 128 | ||
FQ216483.1 | - | 6 - 7 | 743 | 128 | ||
FQ216617.1 | - | 6 - 7 | 746 | 128 | ||
FQ216760.1 | - | 6 - 7 | 744 | 128 | ||
FQ216828.1 | - | 6 - 7 | 743 | 129 | ||
FQ217401.1 | - | 6 - 7 | 744 | 128 | ||
FQ217430.1 | - | 6 - 7 | 744 | 128 | ||
FQ217483.1 | - | 6 - 7 | 747 | 128 | ||
FQ217946.1 | - | 6 - 7 | 745 | 128 | ||
FQ224893.1 | - | 6 - 7 | 746 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
