Assay Details
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024345.2 | NP_001019516.1 | 5 - 6 | 734 | 81 | |
XM_008763681.1 | XP_008761903.1 | 5 - 6 | 1074 | 81 | ||
GenBank mRNA | BC090009.1 | - | 5 - 6 | 655 | 7324151 | 81 |
CF110059.1 | - | 5 - 6 | 620 | 81 | ||
FQ217324.1 | - | 5 - 6 | 671 | 81 | ||
FQ219788.1 | - | 5 - 6 | 631 | 81 | ||
FQ219821.1 | - | 5 - 6 | 671 | 81 | ||
FQ220371.1 | - | 5 - 6 | 671 | 81 | ||
FQ220438.1 | - | 5 - 6 | 631 | 81 | ||
FQ220606.1 | - | 5 - 6 | 671 | 81 | ||
FQ220675.1 | - | 5 - 6 | 671 | 81 | ||
FQ229470.1 | - | 5 - 6 | 671 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
