Assay Details
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC081811.1 | - | 232 | 110 | ||
BC098666.1 | - | 225 | 110 | |||
BC126094.1 | - | 239 | 110 | |||
FQ216025.1 | - | 279 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
