Assay Details
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019187.1 | NP_062060.1 | 3 - 4 | 319 | 72 | |
GenBank mRNA | BC081811.1 | - | 484 | 72 | ||
BC098666.1 | - | 477 | 72 | |||
BC126094.1 | - | 491 | 72 | |||
FQ216025.1 | - | 531 | 72 | |||
FQ230994.1 | - | 1 - 2 | 96 | 72 | ||
L20427.1 | - | 3 - 4 | 319 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
