Assay Details
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013092.2 | NP_001013110.1 | 1 - 2 | 335 | 67 | |
XM_006238050.1 | XP_006238112.1 | 1 - 2 | 361 | 67 | ||
XM_008763672.1 | XP_008761894.1 | 2 - 3 | 165 | 67 | ||
GenBank mRNA | BC062241.1 | - | 1 - 2 | 338 | 6887761 | 67 |
FQ219084.1 | - | 1 - 2 | 313 | 67 | ||
KJ817427.1 | - | 1 - 2 | 179 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
