Assay Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080776.1 | NP_542954.1 | 3 - 4 | 828 | 76 | |
XM_006237921.2 | XP_006237983.1 | 3 - 4 | 894 | 76 | ||
GenBank mRNA | D63886.1 | - | 3 - 4 | 828 | 76 | |
D85509.1 | - | 3 - 4 | 828 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
