Assay Details
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031663.1 | NP_001026833.1 | 5 - 6 | 588 | 89 | |
GenBank mRNA | BC079473.1 | - | 5 - 6 | 594 | 89 | |
BC098713.1 | - | 5 - 6 | 588 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
