Assay Details
Gene Symbol: Hivep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024137.1 | NP_077051.1 | 232 | 145 | ||
GenBank mRNA | D37951.1 | - | 232 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D37951.1, NM_024137.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
