Assay Details
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214458.1 | NP_999623.1 | 10 - 11 | 1631 | 103 | |
XM_008758610.1 | XP_008756832.1 | 11 - 12 | 2788 | 103 | ||
XM_008758611.1 | XP_008756833.1 | 11 - 12 | 2561 | 103 | ||
GenBank mRNA | AY500158.1 | - | 10 - 11 | 1631 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164510 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083439 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083439 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083439 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P185925 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41673 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50206 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37264 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1417 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42187 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25052 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74836 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10716908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716903 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716900 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716906 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716907 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716899 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716904 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716901 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716902 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716909 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716897 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716898 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10716896 | - | - |
Affymetrix Rat Clariom S Assay | PSR0100032721.rn.1 | TC0100004025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032720.rn.1 | TC0100004025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032718.rn.1 | TC0100004025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032719.rn.1 | TC0100004025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032722.rn.1 | TC0100004025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032716.rn.1 | TC0100004025.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10716896 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20847156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350710 | - | - |
RatRef-12 Expression BeadChip | ILMN_54772 | - | - |
