Assay Details
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214458.1 | NP_999623.1 | 2 - 3 | 220 | 154 | |
XM_008758610.1 | XP_008756832.1 | 2 - 3 | 1140 | 154 | ||
XM_008758611.1 | XP_008756833.1 | 2 - 3 | 1138 | 154 | ||
GenBank mRNA | AY500158.1 | - | 2 - 3 | 220 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
