Assay Details
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214458.1 | NP_999623.1 | 3 - 4 | 306 | 125 | |
XM_008758610.1 | XP_008756832.1 | 3 - 4 | 1226 | 125 | ||
XM_008758611.1 | XP_008756833.1 | 3 - 4 | 1224 | 125 | ||
GenBank mRNA | AY500158.1 | - | 3 - 4 | 306 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
