Assay Details
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108457.1 | NP_001101927.1 | 1 - 2 | 104 | 136 | |
XM_006227621.2 | XP_006227683.1 | 1 - 2 | 138 | 136 | ||
GenBank mRNA | FQ225576.1 | - | 1 - 2 | 143 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
