Assay Details
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012062.1 | NP_001012062.1 | 5 - 6 | 1923 | 99 | |
XM_006227620.2 | XP_006227682.1 | 4 - 5 | 1953 | 99 | ||
GenBank mRNA | BC085788.1 | - | 5 - 6 | 1923 | 7108689 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
