Assay Details
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012062.1 | NP_001012062.1 | 6 - 7 | 3106 | 61 | |
XM_006227620.2 | XP_006227682.1 | 6 - 7 | 2146 | 61 | ||
GenBank mRNA | BC085788.1 | - | 6 - 7 | 3106 | 7108689 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P021476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141212 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P021476 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P021476 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P021476 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18039 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18423 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36699 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11545 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60511 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10716662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716663 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10716657 | - | - |
Rat Genome 230 2.0 Array | 1398934_at | - | - |
Rat Genome 230 2.0 Array | 1398934_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398934_at | - | - |
Affymetrix HT Rat Focus Array | 1398934_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100032290.rn.1 | TC0100003957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032292.rn.1 | TC0100003957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032294.rn.1 | TC0100003957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100032291.rn.1 | TC0100003957.rn.1 | RTA TC |
Rat Expression Set 230 | 1398934_PM_at | - | - |
Rat Expression Set 230 | 1398934_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716657 | - | - |
Rat Genome U34 Set | rc_AI060222_at | - | - |
Rat Genome U34 Set | rc_AI137197_at | - | - |
Rat Genome U34 Set | rc_AI178652_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21199435 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376724 | - | - |
