Assay Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107641.1 | NP_001101111.1 | 2 - 3 | 681 | 112 | |
XM_006256604.2 | XP_006256666.1 | 3 - 4 | 969 | 112 | ||
XM_008772994.1 | XP_008771216.1 | 3 - 4 | 809 | 112 | ||
XM_008772995.1 | XP_008771217.1 | 3 - 4 | 2158 | 112 | ||
XM_008772996.1 | XP_008771218.1 | 3 - 4 | 792 | 112 | ||
XM_008772997.1 | XP_008771219.1 | 2 - 3 | 4434 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008772997.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
