Assay Details
Gene Symbol: Prdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772991.1 | XP_008771213.1 | 1 - 2 | 146 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
