Assay Details
Gene Symbol: Prdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107639.2 | NP_001101109.2 | 4 - 5 | 887 | 121 | |
XM_008772991.1 | XP_008771213.1 | 5 - 6 | 852 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
