Assay Details
Gene Symbol: Lin28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001069344.4 | XP_001069344.2 | 3 - 4 | 1807 | 74 | |
XM_006256600.2 | XP_006256662.1 | 3 - 4 | 1241 | 74 | ||
XM_006256601.1 | XP_006256663.1 | 4 - 5 | 672 | 74 | ||
XM_008773029.1 | XP_008771251.1 | 4 - 5 | 1015 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
