Assay Details
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100804.1 | NP_001094274.1 | 3 - 4 | 170 | 125 | |
GenBank mRNA | AY262357.1 | - | 3 - 4 | 159 | 125 | |
BC168196.1 | - | 170 | 7111460 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
