Assay Details
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767042.1 | XP_008765264.1 | 3 - 4 | 578 | 69 | |
GenBank mRNA | AJ272428.1 | - | 3 - 4 | 341 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
