Assay Details
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053495.1 | NP_445947.1 | 2 - 3 | 238 | 64 | |
XM_008767042.1 | XP_008765264.1 | 4 - 5 | 762 | 64 | ||
XM_008767043.1 | XP_008765265.1 | 3 - 4 | 648 | 64 | ||
GenBank mRNA | AB002801.1 | - | 2 - 3 | 238 | 64 | |
AJ272428.1 | - | 4 - 5 | 525 | 64 | ||
AJ272429.1 | - | 3 - 4 | 411 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
