Assay Details
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767043.1 | XP_008765265.1 | 2 - 3 | 466 | 63 | |
XM_008767044.1 | XP_008765266.1 | 2 - 3 | 466 | 63 | ||
GenBank mRNA | AJ272429.1 | - | 2 - 3 | 229 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
