Assay Details
Gene Symbol: Riok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009687.1 | NP_001009687.1 | 7 - 8 | 882 | 69 | |
GenBank mRNA | BC088297.1 | - | 7 - 8 | 882 | 69 | |
FQ232861.1 | - | 7 - 8 | 917 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
