Assay Details
Gene Symbol: LOC688452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135720.1 | NP_001129192.1 | 8 - 9 | 1021 | 90 | |
GenBank mRNA | FM212919.1 | - | 8 - 9 | 1021 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
