Assay Details
Gene Symbol: Fam193b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170408.1 | NP_001163879.1 | 2 - 3 | 173 | 78 | |
XM_006253650.2 | XP_006253712.1 | 1 - 2 | 906 | 78 | ||
XM_006253651.2 | XP_006253713.1 | 2 - 3 | 1210 | 78 | ||
GenBank mRNA | CB719022.1 | - | 2 - 3 | 173 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001170408.1, XM_006253650.2, XM_006253651.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
