Assay Details
Gene Symbol: Fam193b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170408.1 | NP_001163879.1 | 8 - 9 | 2469 | 72 | |
XM_006253646.2 | XP_006253708.1 | 7 - 8 | 2568 | 72 | ||
XM_006253647.2 | XP_006253709.1 | 7 - 8 | 3308 | 72 | ||
XM_006253650.2 | XP_006253712.1 | 7 - 8 | 3202 | 72 | ||
XM_006253651.2 | XP_006253713.1 | 8 - 9 | 3369 | 72 | ||
XM_008771503.1 | XP_008769725.1 | 7 - 8 | 4987 | 72 | ||
GenBank mRNA | BC166819.1 | - | 2 - 3 | 1262 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
